missing translation for 'onlineSavingsMsg'
Learn More

Clofibrate, MedChemExpress

Product Code. 16492119
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492119 1 g 1g
16480858 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492119 Supplier Medchem Express Supplier No. HYB0287
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Clofibrate is an agonist of PPAR, with EC50s of 50 μM, ∼500 μM for murine PPARα and PPARγ, and 55 μM, ∼500 μM for human PPARα and PPARγ, respectively.

Clofibrate is an agonist of PPAR, with EC50s of 50 μM, ∼500 μM for murine PPARα and PPARγ, and 55 μM, ∼500 μM for human PPARα and PPARγ, respectively.

Specifications

Chemical Name or Material Clofibrate
CAS 637-07-0
Color Colorless
Purity Grade Notes Research
Molecular Formula C12H15ClO3
Quantity 1 g
Solubility Information DMSO : ≥ 100 mg/mL (412.03 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC(C)(OC1=CC=C(Cl)C=C1)C(OCC)=O
Molecular Weight (g/mol) 242.7
Formula Weight 242.7
Percent Purity 99.54%
Grade Research
Physical Form Liquid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.