missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Clofazimine

Product Code. 16442246
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442246 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16442246 Supplier MedChemExpress Supplier No. HYB1046
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Clofazimine is an iminophenazine dye, has a marked anti-inflammatory effect, has been used in combination with other antimycobacterial drugs to treat AIDS and Crohn's disease.

Specifications

Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Product Type Dye Reagents
Quantity 500 mg
Shipping Condition Room temperature
CAS 2030-63-9
Molecular Weight (g/mol) 473.4 g/mol
Solubility DMSO : 8.93 mg/mL (18.86 mM; ultrasonic and warming and heat to 60°C) ∣ H2O : < 0.1 mg/mL (insoluble)
Target HBV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.