missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Clofarabine

Product Code. 16414147
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16414147 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16414147 Supplier MedChemExpress Supplier No. HYA0005
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Clofarabine, a nucleoside analogue for research of cancer, is a potent inhibitor of ribonucleotide reductase (IC50=65 nM) by binding to the allosteric site on the regulatory subunit.

Specifications

Molecular Weight (g/mol) 303.68 g/mol
Product Type Reference compound
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Purity 98.05%
Quantity 50 mg
Solubility Information DMSO : ≥ 50 mg/mL (164.65 mM)
For Use With (Application) Cancer-programmed cell death
CAS 123318-82-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.