missing translation for 'onlineSavingsMsg'
Learn More

Clofarabine
GREENER_CHOICE

Product Code. 18738746
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18738746 25 mg 25mg
18777756 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18738746 Supplier Tocris Bioscience Supplier No. 2600/25

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Deoxycytidine kinase (dCK) substrate

Clofarabine is a deoxycytidine kinase (dCK) substrate. Phosphorylated to form clofarabine triphosphate, which competes with dATP for DNA polymerase-α and -ε and potently inhibits ribonucleotide reductase (IC50 = 65 nM). Induces apoptosis by directly altering mitochondrial transmembrane potential. Demonstrates growth inhibition and cytotoxic activity in a variety of leukemias and solid tumors.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloro-9-(2-deoxy-2-fluoro-β-D-arabinofuranosyl)-9H-purin-6-amine
CAS 123318-82-1
Assay Percent Range >99%
Quantity 25 mg
Molecular Formula C10H11ClFN5O3
Formula Weight Observed MW: 303.68
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.