missing translation for 'onlineSavingsMsg'
Learn More

Clodinafop Propargyl, TRC

Product Code. 17974985
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17974985 100 mg 100mg
17983385 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17974985 Supplier Toronto Research Chemicals Supplier No. TRCC584515100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105512-06-9
Molecular Formula C17 H13 Cl F N O4
Molecular Weight (g/mol) 349.74
Synonym Propanoic acid, 2-[4-[(5-chloro-3-fluoro-2-pyridinyl)oxy]phenoxy]-, 2-propynyl ester, (2R)- (9CI), Propanoic acid, 2-[4-[(5-chloro-3-fluoro-2-pyridinyl)oxy]phenoxy]-, 2-propynyl ester, (R)-, (R)-(+)-2-[4-(5-Chloro-3-fluoropyridin-2-yloxy)phenoxy]propionic acid propargyl ester, (R)-2-[4-[(5-Chloro-3-fluoro-2-pyridinyl)oxy]phenoxy]propionic acid 2-propynyl ester, CGA 184927, Clodinafop-propargyl, Prop-2-ynyl (R)-2-[4-(5-chloro-3-fluoropyridin-2-yloxy)phenoxy]propanoate, Topik
IUPAC Name prop-2-ynyl (2R)-2-[4-(5-chloro-3-fluoropyridin-2-yl)oxyphenoxy]propanoate
SMILES C[C@@H](Oc1ccc(Oc2ncc(Cl)cc2F)cc1)C(=O)OCC#C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H13ClFNO4/c1-3-8-22-17(21)11(2)23-13-4-6-14(7-5-13)24-16-15(19)9-12(18)10-20-16/h1,4-7,9-11H,8H2,2H3/t11-/m1/s1
Formula Weight 349.0517
Chemical Name or Material Clodinafop-propargyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.