missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifications
Specifications
| Chemical Name or Material | Clodinafop-propargyl |
| Packaging | Glass Bottle |
| CAS | 105512-06-9 |
| Color | Beige |
| Melting Point | 56°C to 60°C |
| Synonym | 2-propynyl (2R)-2-[4-[(5-chloro-3-fluoro-2-pyridinyl)oxy]phenoxy]propanoate |
| Industry Type | Environmental Analysis |
| Molecular Weight (g/mol) | 349.8 |
| Quantity | 100 mg |
| Grade | Analytical Standard |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?