missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLOCK Rabbit anti-Human, Mouse, Rat, Clone: 3D8H6, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £386.00
Specifications
| Antigen | CLOCK |
|---|---|
| Clone | 3D8H6 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18325284
|
Novus Biologicals
NBP3-16656-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18310605
|
Novus Biologicals
NBP3-16656-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLOCK Monoclonal antibody specifically detects CLOCK in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| CLOCK | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Chromatin Research, Circadian Rhythm, Hypoxia, Neuroendocrinology, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 9575 | |
| IgG | |
| Affinity purified |
| 3D8H6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BHLHE8, circadian locomoter output cycles protein kaput, Class E basic helix-loop-helix protein 8, clock (mouse) homolog, clock homolog (mouse), EC 2.3.1.48, hCLOCK, KAT13D, KIAA0334bHLHe8circadian locomoter output cycles kaput protein | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human CLOCK (O15516). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title