missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLOCK Polyclonal Antibody

Código de producto. 31369225
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity
31369225 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 31369225 Proveedor Invitrogen™ N.º de proveedor PA5150028

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

CLOCK Polyclonal Antibody for Western Blot, ICC/IF, IHC

This gene encodes a protein that belongs to the basic helix-loop-helix (bHLH) family of transcription factors. Polymorphisms within the encoded protein have been associated with circadian rhythm sleep disorders. A similar protein in mice is a circadian regulator that acts as a transcription factor and forms a heterodimer with aryl hydrocarbon receptor nuclear translocator-like to activate transcription of mouse period 1.
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen CLOCK
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene CLOCK
Gene Accession No. O15516
Gene Alias 5330400M04Rik; bHLHe8; circadian locomoter output cycles kaput; circadian locomoter output cycles kaput protein; circadian locomoter output cycles protein kaput; circadian locomotor output cycles kaput; class E basic helix-loop-helix protein 8; CLOCK; clock circadian regulator; clock homolog; hCLOCK; I79_020252; KAT13D; KIAA0334; mCLOCK; rCLOCK
Gene Symbols CLOCK
Host Species Rabbit
Immunogen Recombinant Circadian Locomoter Output Cycles Protein Kaput (CLOCK) (Ala34-Ala379).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9575
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.