missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLOCK Polyclonal Antibody
GREENER_CHOICE

Product Code. 17858522
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17858522 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17858522 Supplier Invitrogen™ Supplier No. PA5140985

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CLOCK Polyclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot

This gene encodes a protein that belongs to the basic helix-loop-helix (bHLH) family of transcription factors. Polymorphisms within the encoded protein have been associated with circadian rhythm sleep disorders. A similar protein in mice is a circadian regulator that acts as a transcription factor and forms a heterodimer with aryl hydrocarbon receptor nuclear translocator-like to activate transcription of mouse period 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLOCK
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CLOCK
Gene Accession No. O08785, O15516, Q9WVS9
Gene Alias 5330400M04Rik; bHLHe8; circadian locomoter output cycles kaput; circadian locomoter output cycles kaput protein; circadian locomoter output cycles protein kaput; circadian locomotor output cycles kaput; class E basic helix-loop-helix protein 8; CLOCK; clock circadian regulator; clock homolog; hCLOCK; I79_020252; KAT13D; KIAA0334; mCLOCK; rCLOCK
Gene Symbols CLOCK
Host Species Rabbit
Immunogen Synthetic peptide within mouse CLOCK 829-855.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12753, 60447, 9575
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.