missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CLOCK Monoclonal antibody specifically detects CLOCK in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | CLOCK |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | HL1099 |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Gene Alias | BHLHE8, circadian locomoter output cycles protein kaput, Class E basic helix-loop-helix protein 8, clock (mouse) homolog, clock homolog (mouse), EC 2.3.1.48, hCLOCK, KAT13D, KIAA0334bHLHe8circadian locomoter output cycles kaput protein |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human CLOCK. The exact sequence is proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?