missing translation for 'onlineSavingsMsg'
Learn More

CLOCK Antibody, Novus Biologicals™

Product Code. 18796793 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18796793 0.025 mL 25µL
18767003 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18796793 Supplier Novus Biologicals Supplier No. NB100126SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 3 publications

CLOCK Polyclonal specifically detects CLOCK in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLOCK
Applications Western Blot, ChIP Assay
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. O08785
Gene Alias BHLHE8, circadian locomoter output cycles protein kaput, Class E basic helix-loop-helix protein 8, clock (mouse) homolog, clock homolog (mouse), EC 2.3.1.48, hCLOCK, KAT13D, KIAA0334bHLHe8circadian locomoter output cycles kaput protein
Gene Symbols CLOCK
Host Species Rabbit
Immunogen Two peptides derived from mouse CLOCK conjugated to albumin. [UniProt# O08785]
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Circadian Rhythm, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 9575
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.