missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Clobenpropit dihydrobromide
GREENER_CHOICE

Product Code. 18798116
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18798116

Brand: Tocris Bioscience™ 0752/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent H3 antagonist and H4 partial agonist

An extremely potent histamine H3 antagonist/inverse agonist (pA2=9.93). Also displays partial agonist activity at H4 receptors; induces eosinophil shape change with an EC50 of 3 nM.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(4-Chlorobenzyl)-S-[3-(4(5)-imidazolyl)propyl]isothiourea dihydrobromide
CAS 145231-35-2
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 100 mM in water
Target Histamine H3 Receptor Antagonists
Molecular Formula C14H17ClN4S0.2 HBr
Formula Weight Observed MW: 470.65
Purity 99%

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.