missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLNS1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38616-100ul
This item is not returnable.
View return policy
Description
CLNS1A Polyclonal antibody specifically detects CLNS1A in Human samples. It is validated for ELISA,Western Blot
Specifications
| CLNS1A | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| chloride channel regulatory protein, Chloride channel, nucleotide sensitive 1A, chloride channel, nucleotide-sensitive, 1A, Chloride conductance regulatory protein ICln, Chloride ion current inducer protein, ClCI, CLNS1B, I(Cln), ICln, methylosome subunit pICln, Reticulocyte pICln, reticulocyte protein ICln | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-237 of human CLNS1A (NP_001284.1).,, Sequence:, MSFLKSFPPPGPAEGLLRQQPDTEAVLNGKGLGTGTLYIAESRLSWLDGSGLGFSLEYPTISLHALSRDRSDCLGEHLYVMVNAKFEEESKEPVADEEEEDSDDDVEPITEFRFVPSDKSALEAMFTAMCECQALHPDPEDEDSDDYDGEEYDVEAHEQGQGDIPTFYTYEEGLSHLTAEGQATLERLEGMLSQSVSSQYNMAGVRTEDSIRDYEDGMEVDTTPTVAGQFEDADVDH | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 1207 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction