missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLN6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584946
This item is not returnable.
View return policy
Description
Immunogen sequence: LVFPLEWFPL NKPSVGDYFH MAYNVITPFL LLKLIERSPR TLPRSIT Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 89%.
CLN6, a 311-amino acid protein, has 7 predicted transmembrane domains and is conserved across vertebrates. The CLN6 protein localizes to the endoplasmic reticulum but contributes to lysosomal function. Mutations in the CLN6 gene cause variant late-onset infantile neuronal ceroid lipofuscinosis (vLINCL), a lysosomal storage disorder marked by progressive mental deterioration and blindness; part of a group of severe inherited neurodegenerative disorders affecting children wherein lysosomes accumulate storage material, causing the death of neurons. CLN6 is one of eight proteins, including CLN1-8, that are associated with NCL.
Specifications
| CLN6 | |
| Polyclonal | |
| Unconjugated | |
| CLN6 | |
| 1810065L06Rik; AW743417; ceroid-lipofuscinosis; ceroid-lipofuscinosis neuronal 6 late infantile; ceroid-lipofuscinosis neuronal protein 6; ceroid-lipofuscinosis, neuronal 6; ceroid-lipofuscinosis, neuronal 6, late infantile, variant; CLN4A; CLN6; D9Bwg1455e; HsT18960; nclf; neuronal ceroid lipofuscinosis; Protein CLN6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54982 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NWW5 | |
| CLN6 | |
| Recombinant protein corresponding to Human CLN6. Recombinant protein control fragment (Product #RP-107980). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction