Learn More
CLN5 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24605-1-AP-20UL
This item is not returnable.
View return policy
Description
The CLN5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CLN5. This antibody recognizes human, mouse antigen. The CLN5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CLN5 | |
| 600 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| CLN5, NCL, Protein CLN5 | |
| 20 μL | |
| 1203 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |