missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLN5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA588837

Product Code. 16556931

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: IPSRRHWPVP YKRFDFRPKP DPYCQAKYTF CPTGSPIPVM EGDDDIEVFR LQAPVWEFKY GDLLGHLKIM HDAIGFRSTL TGKNYTMEWY ELFQLGNCTF PHLRPEMDAP FWCNQGAACF FEGIDDVHWK ENGTLVQVAT ISGNMFNQMA KWVKQDNETG IYYETWNVKA SPEKGAETWF DSYDCSKFVL RTFNKLAEFG AEFKNIETNY TRIFLYSGEP TYLGNETSVF GPTGNKTLGL AIKRFYYPFK PHLPTKEFLL SLLQIFDAVI VHKQFYLFYN FEYWFLPMKF PFIKITYEEI PLPIRNKTLS GL; Positive Samples: Mouse heart, Rat heart; Cellular Location: Lysosome.

This gene is one of eight which have been associated with neuronal ceroid lipofuscinoses (NCL). Also referred to as Batten disease, NCL comprises a class of autosomal recessive, neurodegenerative disorders affecting children. The genes responsible likely encode proteins involved in the degradation of post-translationally modified proteins in lysosomes. The primary defect in NCL disorders is thought to be associated with lysosomal storage function.[provided by RefSeq, Oct 2008].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CLN5
Polyclonal
Unconjugated
CLN5
A730075N08Rik; ceroid-lipofuscinosis; Ceroid-lipofuscinosis neuronal protein 5; ceroid-lipofuscinosis neuronal protein 5 homolog; Ceroid-lipofuscinosis neuronal protein 5 homolog, secreted form; Ceroid-lipofuscinosis neuronal protein 5, secreted form; ceroid-lipofuscinosis, neuronal 5; CLN5; NCL; Protein CLN5
Rabbit
Affinity chromatography
RUO
1203, 211286, 306128
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
2.8 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
O75503, Q3UMW8
CLN5
Recombinant fusion protein containing a sequence corresponding to amino acids 47-358 of human CLN5 (NP_006484.2).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.