missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLN3 Polyclonal Antibody

CLN3 Polyclonal Antibody
Brand: Thermo Scientific PA564103
This item is not returnable.
View return policy
Description
CLN3 Polyclonal Antibody for IHC
Specifications
| CLN3 | |
| Polyclonal | |
| Unconjugated | |
| CLN3 | |
| AI323623; batten disease protein; Battenin; BTS; ceroid lipofuscinosis, neuronal 3, juvenile (Batten, Spielmeyer-Vogt disease); ceroid-lipofuscinosis, neuronal 3; CLN 3; Cln3; CLN3, battenin; Cln3p; JNCL; MGC102840; Protein CLN3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1201 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13286 | |
| CLN3 | |
| Recombinant protein corresponding to Human CLN3. Recombinant protein control fragment (Product #RP-104248). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction