missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102819
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TPP1.
This gene encodes a member of the sedolisin family of serine proteases. The protease functions in the lysosome to cleave N-terminal tripeptides from substrates, and has weaker endopeptidase activity. It is synthesized as a catalytically-inactive enzyme which is activated and auto-proteolyzed upon acidification. Mutations in this gene result in late-infantile neuronal ceroid lipofuscinosis, which is associated with the failure to degrade specific neuropeptides and a subunit of ATP synthase in the lysosome.
Specifications
| CLN2 | |
| Polyclonal | |
| Unconjugated | |
| TPP1 | |
| cell growth-inhibiting gene 1 protein; ceroid-lipofuscinosis, neuronal 2; CLN 2; CLN2; GIG 1; GIG1; growth-inhibiting protein 1; LPIC; lysosomal pepstatin insensitive protease; lysosomal pepstatin-insensitive protease; MGC21297; SCAR7; TPP 1; TPP I; Tpp1; TPP-1; TPPI; TPP-I; Tripeptidyl aminopeptidase; tripeptidyl peptidase 1; tripeptidyl peptidase I; tripeptidyl peptidase-I; tripeptidyl-peptidase 1; Tripeptidyl-peptidase I; UNQ267/PRO304 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1200, 12751, 83534 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14773, O89023, Q9EQV6 | |
| TPP1 | |
| A synthesized peptide derived from human TPP1(Accession O14773), corresponding to amino acid residues S181-L231. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction