missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLK4 Recombinant Human Protein
CLK4 belongs to the CDC2-like protein kinase (CLK) family.
£1772.00 - £11240.00
Specifications
Accession Number | NP_065717 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 85kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15310051
|
Invitrogen™
PR7453A |
100μg |
£1772.00
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10230673
|
Invitrogen™
PV3986 |
1mg |
£11240.00
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CLK4 can interact with and phosphorylate the serine- and arginine-rich (SR) proteins, which are known to play an important role in the formation of spliceosomes, and thus is thought to be involved in the regulation of alternative splicing.
Specifications
NP_065717 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CLK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
85kDa | |
RUO - research use only | |
CLK4 | |
Human | |
GST Tag | |
Frozen |