Learn More
Eurofins DiscoverX CLK3 Protein, Active
Description
CLK3 (CDC Like Kinase 3) is a dual specificity protein kinase which is involved in gene splicing regulation. The CLK family has four family members: CLK1/STY, CLK2, CLK3 and CLK4. CLKs are useful targets for studying diseases attributed to gene mis-splicing events.
GenBank NM_003992. The CLK3 construct contains the translational conflicts T132S and G133S with respect to the accession number given. These are both reported in GB BC019881.
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 63 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥59% |
| Protein | CLK3, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.