missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114721
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CLK3.
CLK3 is a dual specificity kinase that can interact with and cause the nuclear redistribution of SR proteins. Two transcript variants encoding different isoforms have been found for CLK3. Related pseudogenes are located on chromosomes 1 and 9.
Specifications
| CLK3 | |
| Polyclonal | |
| Unconjugated | |
| CLK3 | |
| AI256811; CDC like kinase 3; cdc2/CDC28-like protein kinase 3; CDC-like kinase 3; Clk3; Dual specificity protein kinase CLK3; PHCLK3; PHCLK3/152; Protein kinase CLK3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 102414, 1198, 171305 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O35492, P49761, Q63117 | |
| CLK3 | |
| A synthesized peptide derived from human CLK3(Accession P49761), corresponding to amino acid residues K317-C367. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction