missing translation for 'onlineSavingsMsg'
Learn More

CLK2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-32647

Product Code. 18245960

  • £453.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CLK2 Polyclonal specifically detects CLK2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CLK2
Polyclonal
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
CDC-like kinase 2CLK kinase, clk2, dual specificity protein kinase CLK2, EC 2.7.12.1, hCLK2, MGC61500
Rabbit
60 kDa
100 μL
Protein Kinase
1196
Human
IgG
Western Blot
1.0mg/mL
Western Blot 1:500-1:3000
P49760
CLK2
Recombinant protein encompassing a sequence within the center region of human CLK2. The exact sequence is proprietary.
Affinity Purified
RUO
Primary
Tested in human. Predicted cross-reactivity based on sequence identity: Mouse (98%), Rat (98%). Not yet tested in any other species.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only