missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528653
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (94%), Rat (94%), Xenopus laevis (80%), Pig (97%), Rhesus Monkey (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CLK1 is a dual specificity kinase that phosphorylates PTP-1B and members of the SR family of splicing factors. It is thought to be involved in neuronal differentiation and mRNA splicing. Therefore, the encoded protein may play an indirect role in governing splice site selection.
Specifications
| CLK1 | |
| Polyclonal | |
| Unconjugated | |
| CLK1 | |
| CDC like kinase 1; CDC28/CDC2-like kinase; CDC-like kinase 1; CLK; CLK/STY; CLK1; Dual specificity protein kinase CLK1; Protein kinase CLK1; protein kinase STY; protein tyrosine kinase STY; Sty | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1195 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.17 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P49759 | |
| CLK1 | |
| Recombinant protein encompassing a sequence within the center region of human CLK1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction