Learn More
Thermo Scientific™ ClipTip™ Pipette Tip Racks
GREENER_CHOICE
Product Code. 13256249
Click to view available options
Description:
ClipTip Empty Rack
ClipTip Empty Rack, Large
ClipTip Empty Rack, Medium
ClipTip Empty Rack, Small
Unit Size:
Pack of 10
Pack of 8
Product Code. 13256249
Brand: Thermo Scientific™ 94410219
Only null left
Add to basket
Add to basket
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
  • Small Rack: For use with ClipTip 20 and 50 Pipette Tips
  • Medium Rack: For use with ClipTip 200 and 300 Pipette Tips
  • Large Rack: For use with ClipTip 1000 Pipette Tips
Ordering Information:
Racks are supplied without tips and inserts
TRUSTED_SUSTAINABILITY
Specifications
Description ClipTip Empty Rack, Small
Model ClipTip Empty Rack
For Use With (Equipment) Thermo Scientific E1-ClipTip
Product Line ClipTip