missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLIP4 Polyclonal Antibody

Description
Immunogen sequence: SRVQRVTDSL DTLSEISSNK QNHSYPGFRR SFSTTSASSQ KEINRRNAFS KSKAALRRSW SSTPTAGGIE GSVKLHEGSQ VLLTSSNEMG TV Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 52%.
CLIP4 includes 4 isoforms of the human protein that are produced by alternative splicing.
Specifications
Specifications
| Antigen | CLIP4 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CLIP4 |
| Gene Accession No. | Q8N3C7 |
| Gene Alias | 1700024K14Rik; 1700074B05Rik; 4833417L20Rik; 5830409B12Rik; CAP-Gly domain containing linker protein family member 4; CAP-GLY domain containing linker protein family, member 4; CAP-Gly domain-containing linker protein 4; Clip4; restin-like 2; restin-like protein 2; RGD1311049; RSNL2 |
| Gene Symbols | CLIP4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?