missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLIP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570093
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
Functions as a cytoplasmic linker protein. Involved in TGN-endosome dynamics.
Specifications
| CLIP3 | |
| Polyclonal | |
| Unconjugated | |
| CLIP3 | |
| 1500005P14Rik; AI844915; CAP-GLY domain containing linker protein 3; CAP-Gly domain-containing linker protein 3; clip-170 related protein 59 kDa; CLIP-170-related 59 kDa protein; Clip3; CLIPR59; Clipr-59; Cytoplasmic linker protein 170-related 59 kDa protein; restin-like 1; RGD1306245; RSNL1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 76686 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| B9EHT4 | |
| CLIP3 | |
| synthetic peptide directed towards the middle region of human CLIP3 (aa 9-58). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction