Learn More
Invitrogen™ CLIP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554205
Description
Immunogen sequence: SSSISSVSSV ASSVGGRPSR SGLLTETSSR YARKISGTTA LQEALKEKQQ HIEQLLAERD LERAEVAKAT SHICEVEKEI ALLKAQHEQY VAEAEEKLQR ARLLVESVRK EKVDLSNQLE EERRKVEDLQ FRVEEESITK GDLELTTVA Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.
The protein encoded by this gene belongs to the family of cytoplasmic linker proteins, which have been proposed to mediate the interaction between specific membranous organelles and microtubules. This protein was found to associate with both microtubules and an organelle called the dendritic lamellar body. This gene is hemizygously deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants.
Specifications
| CLIP2 | |
| Polyclonal | |
| Unconjugated | |
| CLIP2 | |
| B230327O20; CAP-Gly domain containing linker protein 2; CAP-Gly domain-containing linker protein 2; CLIP; Clip1; CLIP-115; Clip2; Cyln2; cytoplasmic linker 2; cytoplasmic linker protein 1, 115 kDa; cytoplasmic linker protein 115; Cytoplasmic linker protein 2; Kiaa0291; mKIAA0291; testicular tissue protein Li 40; WBSCR3; WBSCR4; williams-Beuren syndrome chromosomal region 3 protein; Williams-Beuren syndrome chromosomal region 4 protein; Williams-Beuren syndrome chromosome region 3; Williams-Beuren syndrome chromosome region 4; WSCR3; WSCR4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7461 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UDT6 | |
| CLIP2 | |
| Recombinant protein corresponding to Human CLIP2. Recombinant protein control fragment (Product #RP-95914). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.