missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CLIP170 Polyclonal antibody specifically detects CLIP170 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | CLIP170 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 549 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CAP-GLY domain containing linker protein 1, CLIP, CLIP170, CLIP-170Reed-Sternberg intermediate filament-associated protein, CYLN1MGC131604, cytoplasmic linker 1, Cytoplasmic linker protein 1, Cytoplasmic linker protein 170 alpha-2, Reed-Steinberg cell-expressed intermediate filament-associated protein, Restin, restin (Reed-Steinberg cell-expressed intermediate filament-associated protein), RSNCAP-Gly domain-containing linker protein 1 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human CLIP170. The immunogen is located within amino acids 1340 - 1390 of CLIP170. Amino Acid Squence: CNNYDSDDQEKQSKKKPR |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?