missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLIP170 Antibody (CL4851), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61144
This item is not returnable.
View return policy
Description
CLIP170 Monoclonal specifically detects CLIP170 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CLIP170 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| CLIP1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KNLELQLKENKRQLSSSSGNTDTQADEDERAQESQIDFLNSVIVDLQRKNQDLKMKVEMMSEAALNGNGDDLNNYDSDDQEKQSKKKPRLFCDICDCFDLHDTEDCPTQAQMSEDPPHSTHHGSRGEERPYCEICEMFGHWATNCNDDET | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL4851 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:10000 - 1:20000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:10000 - 1:20000 | |
| CAP-GLY domain containing linker protein 1, CLIP, CLIP170, CLIP-170Reed-Sternberg intermediate filament-associated protein, CYLN1MGC131604, cytoplasmic linker 1, Cytoplasmic linker protein 1, Cytoplasmic linker protein 170 alpha-2, Reed-Steinberg cell-expressed intermediate filament-associated protein, Restin, restin (Reed-Steinberg cell-expressed intermediate filament-associated protein), RSNCAP-Gly domain-containing linker protein 1 | |
| Mouse | |
| Protein A purified | |
| Dendritic Cell Markers | |
| 6249 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction