missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLIP170 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76921-0.025mg
This item is not returnable.
View return policy
Description
CLIP170 Polyclonal specifically detects CLIP170 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| CLIP170 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CAP-GLY domain containing linker protein 1, CLIP, CLIP170, CLIP-170Reed-Sternberg intermediate filament-associated protein, CYLN1MGC131604, cytoplasmic linker 1, Cytoplasmic linker protein 1, Cytoplasmic linker protein 170 alpha-2, Reed-Steinberg cell-expressed intermediate filament-associated protein, Restin, restin (Reed-Steinberg cell-expressed intermediate filament-associated protein), RSNCAP-Gly domain-containing linker protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| NP_002947 | |
| CLIP1 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human CLIP170. The immunogen is located within amino acids 1340 - 1390 of CLIP170. Amino Acid Squence: CNNYDSDDQEKQSKKKPR | |
| 0.025 mg | |
| Dendritic Cell Markers | |
| 6249 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction