missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLIP-115/CYLN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25088-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the family of cytoplasmic linker proteins, which have been proposed to mediate the interaction between specific membranous organelles and microtubules. This protein was found to associate with both microtubules and an organelle called the dendritic lamellar body. This gene is hemizygously deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants.Specifications
| CLIP-115/CYLN2 | |
| Polyclonal | |
| Unconjugated | |
| CLIP2 | |
| CLIP, CLIP 115, CLIP2, CYLN2, Cytoplasmic linker protein 115, Cytoplasmic linker protein 2, KIAA0291, WBSCR3, WBSCR4, WSCR3, WSCR4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 269713, 29264, 7461 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O55156, Q9UDT6, Q9Z0H8 | |
| Clip2 | |
| CLIP-115/CYLN2 Fusion Protein Ag18692 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction