missing translation for 'onlineSavingsMsg'
Learn More

Brannan™ Clinical Thermometer with Calibration Certificate

Product Code. 17843018
Change view
Click to view available options
Calibrated:
3 Point Traceable
3-point UKAS
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Calibrated unitSize
17843018 3 Point Traceable Each
17803028 3-point UKAS Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17843018 Supplier Brannan™ Supplier No. 11/064/2 + 49/101/0
Add to basket
Add to basket
This item is not returnable. View return policy

Easy to use waterproof clinical thermometer. Complete with traceable calibration.

  • High accuracy, with fast response
  • Safe, easy to read and simple to use
  • May be used for taking oral, underarm or rectal temperature
  • Temperature range: 32°C to 42.9°C
  • 60 second response time
  • Supplied with calibration
  • Compact, accurate and durable
  • Non-hazardous
  • Complete with battery and instructions

Specifications

Calibration Points 35°C,37°C, 39°C
Certifications/Compliance UK National Standards ISO 9001
For Use With (Application) To take temperature via oral, underarm, or rectal
Length (Metric) 13.3 cm
Material Plastic
Max. Temperature (English) 42.9°F
Max. Temperature (Metric) 42.9°C
Min. Temperature (English) 32°F
Min. Temperature (Metric) 32°C
Temperature Range (English) 32°C to 42.9°C
Temperature Range (Metric) 32°C to 42.9°C
Calibrated 3 Point Traceable
Battery Type 1 x LR41
Accuracy ± 0.1°C between 35.5°C and 42°C, otherwise ± 2°C
Subdivision 0.1°C
Display LCD
Width (Metric) 2.4 cm
IP Rating IP22
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.