missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brannan™ Clinical Thermometer with Calibration Certificate
Click to view available options
Calibrated:
3 Point Traceable
3-point UKAS
Unit Size:
Each
Description
- High accuracy, with fast response
- Safe, easy to read and simple to use
- May be used for taking oral, underarm or rectal temperature
- Temperature range: 32°C to 42.9°C
- 60 second response time
- Supplied with calibration
- Compact, accurate and durable
- Non-hazardous
- Complete with battery and instructions
Specifications
Specifications
| Calibration Points | 35°C, 37°C, 39°C |
| Certifications/Compliance | ISO 17025 UKAS |
| Length (Metric) | 13.3 cm |
| Material | Plastic |
| Max. Temperature (English) | 42.9°C |
| Max. Temperature (Metric) | 42.9°C |
| Min. Temperature (English) | 32°C |
| Min. Temperature (Metric) | 32°C |
| Temperature Range (English) | 32°C to 42.9°C |
| Temperature Range (Metric) | 32°C to 42.9°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction