missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Clinical Chemistry Analyzer Sample Cups

Product Code. 31922887
Change view
Click to view available options
Color:
Blue
Clear
Red
Yellow
Unit Size:
Pack of 12000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
31922887 Red Pack of 12000
31923287 Yellow Pack of 12000
31922808 Blue Pack of 12000
31923264 Clear Pack of 12000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31922887 Supplier Globe Scientific Supplier No. 6500R
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For various laboratory analyzers

Specifications

Closure Type Snap
For Use With (Application) Designed for use on automated instrumentation
For Use With (Equipment) Roche cobas Mira™ analyzer
Color Red
Material Polyethylene
Product Type Sample Cup
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.