missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clindamycin, White Powder, MP Biomedicals™
£87.45 - £431.42
Chemical Identifiers
CAS | 21462-39-5 |
---|---|
Molecular Formula | C18H34Cl2N2O5S |
Molecular Weight (g/mol) | 461.439 |
MDL Number | MFCD07793327 |
InChI Key | AUODDLQVRAJAJM-PIPVPTKYSA-N |
Synonym | (7S)-7-Chloro-7-deoxylincomycin hydrochloride, Cleocin |
PubChem CID | 134129519 |
IUPAC Name | (2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride |
SMILES | CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11466171
|
MP Biomedicals
0219116810 |
10 mg |
£87.45
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11476171
|
MP Biomedicals
0219116850 |
50 mg |
£240.62
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11486171
|
MP Biomedicals
0219116880 |
100 mg |
£431.42
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
21462-39-5 | |
461.439 | |
AUODDLQVRAJAJM-PIPVPTKYSA-N | |
134129519 | |
CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl |
C18H34Cl2N2O5S | |
MFCD07793327 | |
(7S)-7-Chloro-7-deoxylincomycin hydrochloride, Cleocin | |
(2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride |
Specifications
Clindamycin | |
21462-39-5 | |
MFCD07793327 | |
AUODDLQVRAJAJM-PIPVPTKYSA-N | |
(2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride | |
134129519 | |
White |
Solid | |
C18H34Cl2N2O5S | |
(7S)-7-Chloro-7-deoxylincomycin hydrochloride, Cleocin | |
CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl | |
461.439 | |
461.4 | |
Clindamycin |
Safety and Handling
Recommended Storage : Store at 4°C.
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Clindamycin Hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : clindamycim hydrochloride