missing translation for 'onlineSavingsMsg'
Learn More

Clindamycin phosphate, MedChemExpress

Product Code. 16428586
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16428586

Brand: Medchem Express HYB1064

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Clindamycin phosphate (Clindamycin 2-phosphate) is a broad-spectrum bacteriostatic lincosamide antibiotic. Clindamycin phosphate is the prodrug of Clindamycin (HY-B1455) with no antimicrobial activity in vitro but can be rapidly converted in vivo to the active parent drug, Clindamycin, by phosphatase ester hydrolysis. Clindamycin phosphate can be used for researching acne and bacterial vaginosis.

Clindamycin phosphate (Clindamycin 2-phosphate) is a broad-spectrum bacteriostatic lincosamide antibiotic. Clindamycin phosphate is the prodrug of Clindamycin (HY-B1455) with no antimicrobial activity in vitro but can be rapidly converted in vivo to the active parent drug, Clindamycin, by phosphatase ester hydrolysis. Clindamycin phosphate can be used for researching acne and bacterial vaginosis.

Specifications

Chemical Name or Material Clindamycin phosphate
Physical Form Solid
CAS 24729-96-2
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H34ClN2O8PS
For Use With (Application) Neuroscience-Neuromodulation
Synonym Clindamycin 2-phosphate U-28508
Solubility Information H2O : ≥ 320 mg/mL (633.71 mM)
SMILES CCC[C@H]1CN(C)[C@H](C(N[C@@]([H])([C@@]2([H])O[C@H](SC)[C@H](OP(O)(O)=O)[C@@H](O)[C@H]2O)[C@@H](Cl)C)=O)C1
Molecular Weight (g/mol) 504.96
Formula Weight 504.96
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.