missing translation for 'onlineSavingsMsg'
Learn More

Clindamycin Palmitate Hydrochloride, TRC

Product Code. 30360111
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30360111 1 g 1g
30351913 100 mg 100mg
30354386 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30360111 Supplier Toronto Research Chemicals Supplier No. TRCC5800351G
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25507-04-4
Molecular Formula C34 H63 Cl N2 O6 S . Cl H
Molecular Weight (g/mol) 699.85
Synonym Clindamycin Palmitate Hydrochloride, L-threo-α-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-, 2-hexadecanoate, monohydrochloride, L-threo-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-(1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-,2-palmitate, monohydrochloride, trans-α-, L-threo-α-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-[[(1-methyl-4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-, 2-hexadecanoate, monohydrochloride, (2S-trans)-, Palmitic acid, 2-ester with methyl 7-chloro-6,7,8-trideoxy-6-(trans-1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-L-threo-α-D-galacto-octopyranoside monohydrochloride, Cleocin Pediatric, Clindamycin 2-palmitate hydrochloride, Clindamycin palmitate hydrochloride
IUPAC Name [(2R,3R,4S,5R,6R)-6-[(1S,2S)-2-chloro-1-[[(2S,4R)-1-methyl-4-propylpyrrolidine-2-carbonyl]amino]propyl]-4,5-dihydroxy-2-methylsulfanyloxan-3-yl] hexadecanoate;hydrochloride
SMILES Cl.CCCCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](O)[C@@H](O)[C@H](O[C@@H]1SC)[C@H](NC(=O)[C@@H]2C[C@@H](CCC)CN2C)[C@H](C)Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C34H63ClN2O6S.ClH/c1-6-8-9-10-11-12-13-14-15-16-17-18-19-21-27(38)42-32-30(40)29(39)31(43-34(32)44-5)28(24(3)35)36-33(41)26-22-25(20-7-2)23-37(26)4;/h24-26,28-32,34,39-40H,6-23H2,1-5H3,(H,36,41);1H/t24-,25+,26-,28+,29+,30-,31+,32+,34+;/m0./s1
Formula Weight 698.3862
Chemical Name or Material Clindamycin Palmitate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.