missing translation for 'onlineSavingsMsg'
Learn More

Clindamycin hydrochloride monohydrate

Product Code. 15484009
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15484009 5 g 5g
15474009 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484009 Supplier Thermo Scientific Alfa Aesar Supplier No. J61409.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Clindamycin hydrochloride, CAS # 21462-39-5, is the salt of clindamycin, a derivative of the broad-spectrum bacteriostatic antibiotic lincomycin. | CAS: 58207-19-5 | C18H34Cl2N2O5S | 461.439 g/mol

Clindamycin hydrochloride monohydrate is an antibacterial agent that is a semisynthetic analog of lincomycin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Clindamycin is a lincosamide antibiotic that disrupts protein synthesis by interfering with the aminoacyl-tRNA ribosome complex

• Clindamycin decreases sebaceous gland activity and blocks the neuromuscular junction

Application

• Clindamycin is commonly used in laboratory applications to inhibit the growth anaerobic microbes, but has some effectiveness against aerobic microbes as well

• It can be used in the laboratory study of cell culture for resistance testing, or in antibiotic allergic reaction studies

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58207-19-5
Molecular Formula C18H34Cl2N2O5S
Molecular Weight (g/mol) 461.439
MDL Number MFCD07793327
InChI Key AUODDLQVRAJAJM-NFOLQJLNSA-N
PubChem CID 131632848
IUPAC Name (2S,4R)-N-[2-chloro-1-[(2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
SMILES CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl

Specifications

Type Clindamycin Hydrochloride Monohydrate
Physical Form Powder
Color White
Quantity 5 g
Beilstein 4070786
Merck Index 14,2356
Solubility Information Soluble in water
Formula Weight 479.46 (461.44 Anhydrous)
Chemical Name or Material Clindamycin hydrochloride monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.