missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clindamycin Hydrochloride, Mikromol™
Description
Clindamycin Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Clindamycin hydrochloride |
| CAS | 21462-39-5 |
| Analyte or Component Names | Clindamycin hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H33 Cl N2 O5 S . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Clindamycin Hydrochloride, 7(S)-Chloro-7-deoxylincomycin hydrochloride, 7-Chloro-7-deoxylincomycin hydrochloride, Antirobe vet., Cleocin hydrochloride, Clindamycin hydrochloride, Clindavet, Clindimycin hydrochloride, Clindomycin hydrochloride, Dalacina, Methyl 7-Chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-L-threo-alpha-D-galacto-octopyranoside hydrochloride (1:1), trans-alpha-Methyl 7-Chloro-6,7,8-trideoxy-6-(1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-L-threo-alpha-D-galacto-octopyranoside monohydrochloride, (2S-trans)-Methyl 7-Chloro-6,7,8-trideoxy-6-[[(1-methyl-4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-L-threo-alpha-D-galacto-octopyranoside monohydrochloride, Methyl 7-Chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-L-threo-alpha-D-galacto-octopyranoside monohydrochloride |
| InChI Formula | InChI=1S/C18H33ClN2O5S.ClH/c1-5-6-10-7-11(21(3)8-10)17(25)20-12(9(2)19)16-14(23)13(22)15(24)18(26-16)27-4;/h9-16,18,22-24H,5-8H2,1-4H3,(H,20,25);1H/t9-,10+,11-,12+,13-,14+,15+,16+,18+;/m0./s1 |
| SMILES | Cl.CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@H](C)Cl)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O |
| IUPAC Name | (2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?