missing translation for 'onlineSavingsMsg'
Learn More

Clindamycin hydrochloride, MedChemExpress

Product Code. 16482159
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482159 1 g 1g
16458286 100 mg 100mg
16450918 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16482159 Supplier Medchem Express Supplier No. HYB0408A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Clindamycin (hydrochloride) is a semisynthetic lincosamide antibiotic, which inhibits protein synthesis by acting on the 50S ribosomal.

Clindamycin (hydrochloride) is a semisynthetic lincosamide antibiotic, which inhibits protein synthesis by acting on the 50S ribosomal.

Specifications

Chemical Name or Material Clindamycin hydrochloride
Physical Form Solid
CAS 21462-39-5
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C18H34Cl2N2O5S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (216.71 mM) ∣H2O : ≥ 100 mg/mL (216.71 mM)
SMILES C[C@H](Cl)[C@@]([C@@]([C@@H]([C@H](O)[C@H]1O)O)([H])O[C@@H]1SC)([H])NC([C@@H]2C[C@@H](CCC)CN2C)=O.Cl
Molecular Weight (g/mol) 461.44
Formula Weight 461.44
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.