missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clindamycin 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Clindamycin hydrochloride |
| CAS | 21462-39-5 |
| Analyte or Component Names | Clindamycin hydrochloride |
| Shipping Condition | +20°C |
| Molecular Formula | C18 H33 Cl N2 O5 S . Cl H |
| Concentration | 1000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Clindamycin Hydrochloride, 7(S)-Chloro-7-deoxylincomycin hydrochloride, 7-Chloro-7-deoxylincomycin hydrochloride, Antirobe vet., Cleocin hydrochloride, Clindamycin hydrochloride, Clindavet, Clindimycin hydrochloride, Clindomycin hydrochloride, Dalacina, Methyl 7-Chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-L-threo-alpha-D-galacto-octopyranoside hydrochloride (1:1), trans-alpha-Methyl 7-Chloro-6,7,8-trideoxy-6-(1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-L-threo-alpha-D-galacto-octopyranoside monohydrochloride, (2S-trans)-Methyl 7-Chloro-6,7,8-trideoxy-6-[[(1-methyl-4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-L-threo-alpha-D-galacto-octopyranoside monohydrochloride, Methyl 7-Chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-L-threo-alpha-D-galacto-octopyranoside monohydrochloride |
| InChI Formula | InChI=1S/C18H33ClN2O5S.ClH/c1-5-6-10-7-11(21(3)8-10)17(25)20-12(9(2)19)16-14(23)13(22)15(24)18(26-16)27-4;/h9-16,18,22-24H,5-8H2,1-4H3,(H,20,25);1H/t9-,10+,11-,12+,13-,14+,15+,16+,18+;/m0./s1 |
| SMILES | Cl.CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@H](C)Cl)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?