missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clinafloxacin, MedChemExpress
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Clinafloxacin (AM 1091) is a potent and broad-spectrum fluoroquinolone antibiotic, has inhibitory activity against gram-positive, gram-negative bacterias, and anaerobic pathogens in vitro. Clinafloxacin is against DNA gyrase and topoisomerase IV of S. aureus with IC50 values of 0.92 μg/ml and 1.62 μg/ml, respectively.
Specifications
Specifications
| Chemical Name or Material | Clinafloxacin |
| Physical Form | Powder |
| CAS | 105956-97-6 |
| Color | Brown |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C17H17ClFN3O3 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | AM-1091 CI-960 PD 127391 |
| Solubility Information | DMSO : 2 mg/mL (5.47 mM; ultrasonic and warming and heat to 80°C) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction