missing translation for 'onlineSavingsMsg'
Learn More

Clinafloxacin Hydrochloride, TRC

Product Code. 30135774
Change view
Click to view available options
Quantity:
10 mg
250 mg
1 g
Unit Size:
10mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135774 250 mg 250mg
30136822 1 g 1g
30005632 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135774 Supplier Toronto Research Chemicals Supplier No. TRCC579000250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105956-99-8
Molecular Formula C17 H17 Cl F N3 O3 . Cl H
Molecular Weight (g/mol) 402.25
Synonym Clinafloxacin Hydrochloride, 3-Quinolinecarboxylic acid, 7-(3-amino-1-pyrrolidinyl)-8-chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-, monohydrochloride (9CI), AM 1091, CI 960, Clinafloxacin hydrochloride, KR 11095, 3-Quinolinecarboxylic acid, 7-(3-amino-1-pyrrolidinyl)-8-chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-, hydrochloride (1:1)
IUPAC Name 7-(3-aminopyrrolidin-1-yl)-8-chloro-1-cyclopropyl-6-fluoro-4-oxoquinoline-3-carboxylic acid;hydrochloride
SMILES Cl.NC1CCN(C1)c2c(F)cc3C(=O)C(=CN(C4CC4)c3c2Cl)C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H17ClFN3O3.ClH/c18-13-14-10(5-12(19)15(13)21-4-3-8(20)6-21)16(23)11(17(24)25)7-22(14)9-1-2-9;/h5,7-9H,1-4,6,20H2,(H,24,25);1H
Formula Weight 401.0709
Chemical Name or Material Clinafloxacin Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.