Learn More
Nickel Electro™ Clifton™ Stainless Steel Student Spirometer
Nickel Electro™ Clifton™ Stainless Steel Student Spirometer can be used for investigating tidal volumes, respiratory rate, pulmonary ventilation, reserve volumes and vital capacity.
Brand: Nickel Electro™ SM-1
This item is not returnable.
View return policy
Description
- A simple lever control enables the subject to be connected either to atmosphere or to the float for recording.
- A container for soda-lime crystals is fitted which can be used to absorb carbon dioxide.
- The spirometer is designed to offer minimum resistance to air flow.
- This means greater comfort for the subject and more realistic results.
- The tank is one piece stainless steel pressing housed in a durable painted outer case.
- The float is constructed from lightweight plastic.
- The float is calibrated 0 to 7 x 0.5 litres and has a metal counterpoise.
- The pen arm corresponds to approximately 0.5 litres.
- Supplied with a mouth piece that fits into the corrugated breathing hoses which snugly connects into Spirometer.
- Also includes a nose clip and pen arm for fitting data logger sensors.
Specifications
| Included a nose clip and pen arm for fitting data logger sensors. | |
| 1/Ea. | |
| Stainlea Steel, Plastic |