Learn More
Nickel Electro™ Clifton™ Stainless Steel Raised Shelf
Click to view available options
Size:
30 x 30 x 20 cm
30 x 35 x 20 cm
60 x 35 x 20 cm
For Use With (Equipment):
14, 22, 28 And 38 Liter Baths
38 Liter Baths
NE4-22T, NE4-28T, NE4-28D, NE4-22E
Unit Size:
Each
Description
- Simple solution for placing shallow vessels in a stirred bath
- Manufactured from stainless steel
- The raised shelf is perforated to allow free circulation of water
- Can be easily removed and cleaned
- Shelf can be inverted to provide two different immersion depths
Specifications
Specifications
| For Use With (Equipment) | NE4-22T, NE4-28T, NE4-28D, NE4-22E |
| Type | Shelf Raised |
| Material | Stainless Steel |
| Size | 30 x 35 x 20 cm |