Learn More
Nickel Electro™ Clifton™ Stainless Steel Raised Shelf
Description
- Simple solution for placing shallow vessels in a stirred bath
- Manufactured from stainless steel
- The raised shelf is perforated to allow free circulation of water
- Can be easily removed and cleaned
- Shelf can be inverted to provide two different immersion depths
Specifications
Specifications
| For Use With (Equipment) | 14, 22, 28 And 38 Liter Baths |
| Type | Shelf Raised |
| Material | Stainless Steel |
| Size | 30 x 30 x 20 cm |
| Dimensions (L x W x H) | 30 x 30 x 20 cm |