Description
- Stainless steel
- Insulated
- Minimises heat loss
- Reduces evaporation
- Assists in maintaining bath temperature
Specifications
Specifications
| For Use With (Equipment) | NE1, NE2, NE3 series 14 L Baths |
| Material | Stainless Steel |