Description
- Stainless steel
- Insulated
- Minimises heat loss
- Reduces evaporation
- Assists in maintaining bath temperature
Specifications
Specifications
| For Use With (Equipment) | Nickel-Electro NE4 series 8L Baths |
| Material | Stainless Steel |