missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Electro™ Clifton™ Stainless Steel Flat Taper Blade
Description
- Quick to clean and has an attractive shiny appearance making it the material of choice for applications requiring sterile surfaces.
- Immune to sterilizing solutions, most of the organic chemicals and dyestuffs, and a wide variety of inorganic chemicals
- Non-magnetic
- Bright finish
Specifications
Specifications
| Quantity | 5 Pk. |
| Product Type | Chattaway Double Spatula |
| Width (Metric) | 10 mm |
| Dimensions (L x W) | 180 x 10 mm |
| Material | Nickel |
| Length (Metric) | 180 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?