missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLIC4 Recombinant Rabbit Monoclonal Antibody (23GB3465), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553416
This item is not returnable.
View return policy
Description
Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 4 (CLIC4) protein, encoded by the CLIC4 gene, is a member of the p64 family; the gene is expressed in many tissues and exhibits a intracellular vesicular pattern in Panc-1 cells (pancreatic cancer cells).
Specifications
| CLIC4 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9QYB1, Q9Y696, Q9Z0W7 | |
| CLIC4 | |
| A synthetic peptide of human CLIC4 (1-28AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3465 | |
| Unconjugated | |
| CLIC4 | |
| chloride intracellular channel 4; chloride intracellular channel 4 (mitochondrial); chloride intracellular channel 4 like; chloride intracellular channel protein 4; Clic4; CLIC4L; D0Jmb3; DKFZP566G223; FLJ38640; H1; huH1; intracellular chloride ion channel protein p64H1; mc3s5; mc3s5/mtCLIC; mitochondrial chloride intracellular channel 4; MTCLIC; p64H1; TU-74 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25932, 29876, 83718 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction